Läs mer
1-Cyclopropyl-2-(triphenylphosphoranylidene)-ethanone, TRC
Produktkod. 30363010
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30363010 100 mg
1 options
Produktkod. 30363010 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC988870100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Cyclopropyl-2-(triphenylphosphoranylidene)-ethanone
CAS 7691-76-1
Molekylformel C23H21OP
Kvantitet 100 mg
Synonym Cyclopropyl (Triphenylphosphoranylidene)methyl Ketone, [(Cyclopropylcarbonyl)methylene]triphenyl-phosphorane
InChI formel InChI=1S/C23H21OP/c24-23(19-16-17-19)18-25(20-10-4-1-5-11-20,21-12-6-2-7-13-21)22-14-8-3-9-15-22/h1-15,18-19H,16-17H2
LEDER O=C(C=P(c1ccccc1)(c2ccccc2)c3ccccc3)C4CC4
IUPAC-namn 1-cyclopropyl-2-(triphenyl-λ^{5}-phosphanylidene)ethanone
Molekylvikt (g/mol) 344.39
Formel vikt 344.133