Läs mer
1-Cyclopropyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid, Mikromol™
Produktkod. 30458233
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30458233 4 x 25 mg
30051236 100 mg
30050933 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30458233 Leverantör Mikromol Leverantörsnummer MM0885.1400254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Cyclopropyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Cyclopropyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid
CAS 94695-52-0
Analyt- eller komponentnamn 1-Cyclopropyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid
Frakt skick Room Temperature
Molekylformel C13 H8 F3 N O3
Certifikat och efterlevnad ISO 17025
Synonym 3-Quinolinecarboxylic acid, 1-cyclopropyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-, 1-Cyclopropyl-1,4-dihydro-6,7,8-trifluoro-4-oxoquinoline-3-carboxylic acid, 1-Cyclopropyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid, 1-Cyclopropyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinolin-3-carboxylic acid, 1-Cyclopropyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
InChI formel InChI=1S/C13H8F3NO3/c14-8-3-6-11(10(16)9(8)15)17(5-1-2-5)4-7(12(6)18)13(19)20/h3-5H,1-2H2,(H,19,20)
Lösningstyp Neat
LEDER OC(=O)C1=CN(C2CC2)c3c(F)c(F)c(F)cc3C1=O
IUPAC-namn 1-cyclopropyl-6,7,8-trifluoro-4-oxoquinoline-3-carboxylic acid
Molekylvikt (g/mol) 283.2
Kvantitet 4 x 25 mg
Formel vikt 283.0456 g/mol