Läs mer
1-Cyclopropyl-8-ethoxy-6,7-difluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester, TRC
Produktkod. 30366277
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30366277 5 mg
1 options
Produktkod. 30366277 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9885055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Cyclopropyl-8-ethoxy-6,7-difluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester
CAS 172602-83-4
Molekylformel C17H17F2NO4
Kvantitet 5 mg
Synonym 3-Quinolinecarboxylic acid, 1-cyclopropyl-8-ethoxy-6,7-difluoro-1,4-dihydro-4-oxo-, ethyl ester, Ethyl 1-cyclopropyl-8-ethoxy-6,7-difluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate
InChI formel InChI=1S/C17H17F2NO4/c1-3-23-16-13(19)12(18)7-10-14(16)20(9-5-6-9)8-11(15(10)21)17(22)24-4-2/h7-9H,3-6H2,1-2H3
LEDER CCOC(=O)C1=CN(C2CC2)c3c(OCC)c(F)c(F)cc3C1=O
IUPAC-namn ethyl 1-cyclopropyl-8-ethoxy-6,7-difluoro-4-oxoquinoline-3-carboxylate
Molekylvikt (g/mol) 337.32
Formel vikt 337.1126