Learn More
BRAND™ 1 D Filter Crucible
Filter Crucible constructed of clear glass for visibility.
3565.00 SEK
Specifications
Diameter (Metric) Disc | 3 cm |
---|---|
Material | Borosilicate Glass |
Capacity (Metric) | 30 mL |
Product Type Specs | Crucible |
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
15282439
|
BRAND™
458021 |
1 |
3565.00 SEK
Pack of 10 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
15292439
|
BRAND™
458022 |
2 |
3565.00 SEK
Pack of 10 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
15202449
|
BRAND™
458023 |
3 |
3565.00 SEK
Pack of 10 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
Description
- Constructed of Borosilicate Glass 3.3
Specifications
3 cm | |
Borosilicate Glass | |
30 mL | |
Crucible |