Läs mer
1-Deoxy-D-xylulose (Aqueous Solution, 1 M), TRC
Produktkod. 30348979
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30348979 5 mg
30364879 10 mg
30366657 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30348979 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2818805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 60299-43-6
Molekylformel C5H10O4
Molekylvikt (g/mol) 134.13
Synonym 1-Deoxy-D-threo-2-pentulose, 1-Deoxy-D-threo-pentulose, 1-Deoxy-D-xylulose, D-1-Deoxyxylulose
IUPAC-namn (3S,4R)-3,4,5-trihydroxypentan-2-one
LEDER CC(=O)[C@@H](O)[C@H](O)CO
Specifikationer
Kemiskt namn eller material 1-Deoxy-D-xylulose
Kvantitet 5 mg
InChI formel InChI=1S/C5H10O4/c1-3(7)5(9)4(8)2-6/h4-6,8-9H,2H2,1H3/t4-,5-/m1/s1
Formel vikt 134.0579