missing translation for 'onlineSavingsMsg'
Läs mer

1-Deoxy-D-xylulose (Aqueous Solution, 1 M), TRC

Produktkod. 30366657
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366657 50 mg
30348979 5 mg
30364879 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30366657 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28188050MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 60299-43-6
Molekylformel C5H10O4
Molekylvikt (g/mol) 134.13
Synonym 1-Deoxy-D-threo-2-pentulose, 1-Deoxy-D-threo-pentulose, 1-Deoxy-D-xylulose, D-1-Deoxyxylulose
IUPAC-namn (3S,4R)-3,4,5-trihydroxypentan-2-one
LEDER CC(=O)[C@@H](O)[C@H](O)CO

Specifikationer

Kemiskt namn eller material 1-Deoxy-D-xylulose
Kvantitet 50 mg
InChI formel InChI=1S/C5H10O4/c1-3(7)5(9)4(8)2-6/h4-6,8-9H,2H2,1H3/t4-,5-/m1/s1
Formel vikt 134.0579
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.