Läs mer
1-Deoxynojirimycin, MedChemExpress
Produktkod. 16473446
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16473446 25 mg
16458807 5 mg
16422347 50 mg
16416218 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16473446 Leverantör Medchem Express Leverantörsnummer HY14860
MedChemExpress 1-Deoxynojirimycin (Duvoglustat) is a potent and orally active α-glucosidase inhibitor. 1-Deoxynojirimycin suppresses postprandial blood glucose and is widely used for diabetes mellitus. 1-Deoxynojirimycin possesses antihyperglycemic, anti-obesity, and antiviral features.
1-Deoxynojirimycin (Duvoglustat) is a potent and orally active α-glucosidase inhibitor. 1-Deoxynojirimycin suppresses postprandial blood glucose and is widely used for diabetes mellitus. 1-Deoxynojirimycin possesses antihyperglycemic, anti-obesity, and antiviral features.
Specifikationer
Kemiskt namn eller material 1-Deoxynojirimycin
Fysisk form Solid
CAS 19130-96-2
Färg White
Kvantitet 25 mg
Anteckningar om renhetsgrad Research
Molekylformel C6H13NO4
För användning med (applikation) Metabolism-sugar/lipid metabolism
Synonym Duvoglustat
Löslighetsinformation H2O : ≥ 34 mg/mL (208.37 mM)
LEDER O[C@@H]1[C@@H](CO)NC[C@H](O)[C@H]1O
Molekylvikt (g/mol) 163.17
Formel vikt 163.17
Procent renhet 98.0%