Läs mer
1-Descarboxy Ketorolac, TRC
Produktkod. 30136631
Change view
Kvantitet:
2.5 mg
20 mg
50 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30136631 50 mg -
30132926 20 mg -
17921225 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136631 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28943550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 113502-55-9
Molekylformel C14 H13 N O
Molekylvikt (g/mol) 211.26
Synonym Phenyl(2,3-dihydro-1H-pyrrolizin-5-yl)methanone, Ketorolac Trometamol Imp. I (EP)
IUPAC-namn 6,7-dihydro-5H-pyrrolizin-3-yl(phenyl)methanone
LEDER O=C(c1ccccc1)c2ccc3CCCn23
Specifikationer
Kemiskt namn eller material Phenyl(2,3-dihydro-1H-pyrrolizin-5-yl)methanone
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C14H13NO/c16-14(11-5-2-1-3-6-11)13-9-8-12-7-4-10-15(12)13/h1-3,5-6,8-9H,4,7,10H2
Formel vikt 211.0997
Procent renhet >95%