missing translation for 'onlineSavingsMsg'
Läs mer

1-Desoxycarbadox, TRC

Produktkod. 30362007
Change view
Click to view available options
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30362007 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30362007 Supplier Toronto Research Chemicals Supplier No. TRCD296999100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 1-Desoxycarbadox
CAS 118828-83-4
Molekylformel C11 H10 N4 O3
Kvantitet 100 mg
Synonym (E)-3-((2-(Methoxycarbonyl)hydrazono)methyl)quinoxaline1-Oxide
InChI formel InChI=1S/C11H10N4O3/c1-18-11(16)14-12-6-8-7-15(17)10-5-3-2-4-9(10)13-8/h2-7H,1H3,(H,14,16)
LEDER COC(=O)NN=Cc1cn(=O)c2ccccc2n1
IUPAC-namn methyl N-[(4-oxoquinoxalin-2-yl)methylideneamino]carbamate
Molekylvikt (g/mol) 246.222
Formel vikt 246.075
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.