missing translation for 'onlineSavingsMsg'
Learn More

(1-Dodecyl)trimethylammonium chloride, 97%

Product Code. 11449046
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 112-00-5 | C15H34ClN | 263.894 g/mol

Dodecyltrimethylammonium Chloride is useful as a paint stripper, a foaming stabilizer, and a bactericidal lotion. It is used in coating, plastics, paints, rubber and ink production, as anti-static agent, catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecyltrimethylammonium Chloride is useful as a paint stripper, a foaming stabilizer, and a bactericidal lotion. It is used in coating, plastics, paints, rubber and ink production, as anti-static agent, catalyst.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-00-5
Molecular Formula C15H34ClN
Molecular Weight (g/mol) 263.894
MDL Number MFCD00041974
InChI Key DDXLVDQZPFLQMZ-UHFFFAOYSA-M
Synonym dodecyltrimethylammonium chloride, alicop, cation bb, aliquat 4, n,n,n-trimethyldodecan-1-aminium chloride, laurtrimonium chloride, dehyquart lt, cation fb, nissan cation bb, lauryltrimethylammonium chloride
PubChem CID 8152
IUPAC Name dodecyl(trimethyl)azanium;chloride
SMILES CCCCCCCCCCCC[N+](C)(C)C.[Cl-]

Specifications

Melting Point ∼237°C (decomposition)
Odor Odorless
Linear Formula CH3(CH2)11N(CH3)3Cl
Quantity 25 g
UN Number UN3077
Beilstein 3915951
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 263.9
Percent Purity 97%
Chemical Name or Material (1-Dodecyl)trimethylammonium chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.