Läs mer
1'-Epi 2',2'-Difluoro-2'-deoxyuridine, TRC
Produktkod. 17933045
Change view
Kvantitet:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17933045 10 mg
17911445 1 mg
17923045 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17933045 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE58480010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 153381-14-7
Molekylformel C9H10F2N2O5
Molekylvikt (g/mol) 264.18
Synonym 1-(2-Deoxy-2,2-difluoro-α-D-erythro-pentofuranosyl)-2,4(1H,3H)-pyrimidinedione, 1'-Epi-2'-deoxy-2',2'-difluorouridine;
LEDER C1=CN(C(=O)NC1=O)[C@@H]2C([C@H]([C@H](O2)CO)O)(F)F
Specifikationer
Kemiskt namn eller material 1'-Epi 2',2'-Difluoro-2'-deoxyuridine
Kvantitet 10 mg
InChI formel InChI=1S/C9H10F2N2O5/c10-9(11)6(16)4(3-14)18-7(9)13-2-1-5(15)12-8(13)17/h1-2,4,6-7,14,16H,3H2,(H,12,15,17)/t4-,6+,7+/m1/s1