Läs mer
1'-Epi Gemcitabine Hydrochloride, TRC
Produktkod. 30012892
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012892 50 mg
17963245 10 mg
17981865 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012892 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE58851050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 122111-05-1
Molekylformel C9H12ClF2N3O4
Molekylvikt (g/mol) 299.66
Synonym 2(1H)-Pyrimidinone, 4-amino-1-(2-deoxy-2,2-difluoro-α-D-erythro-pentofuranosyl)-, hydrochloride (1:1), 2(1H)-Pyrimidinone, 4-amino-1-(2-deoxy-2,2-difluoro-α-D-erythro-pentofuranosyl)-, monohydrochloride (9CI), 1'-epi-Gemcitabine hydrochloride
LEDER Cl.NC1=NC(=O)N(C=C1)[C@H]2O[C@H](CO)[C@@H](O)C2(F)F
Specifikationer
Kemiskt namn eller material 1'-Epi Gemcitabine Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C9H11F2N3O4.ClH/c10-9(11)6(16)4(3-15)18-7(9)14-2-1-5(12)13-8(14)17;/h1-2,4,6-7,15-16H,3H2,(H2,12,13,17);1H/t4-,6-,7+;/m1./s1
Formel vikt 299.0484
Procent renhet >95%