Läs mer
(1-Ethoxyvinyl)trimethylsilane, TRC
Produktkod. 30179367
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179367 100 mg
30179014 500 mg
30179572 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179367 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE937906100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 81177-92-6
Molekylformel C7H16OSi
Molekylvikt (g/mol) 144.29
IUPAC-namn 1-ethoxyethenyl(trimethyl)silane
LEDER CCOC(=C)[Si](C)(C)C
Specifikationer
Kemiskt namn eller material (1-Ethoxyvinyl)trimethylsilane
Kvantitet 100 mg
InChI formel InChI=1S/C7H16OSi/c1-6-8-7(2)9(3,4)5/h2,6H2,1,3-5H3
Formel vikt 144.097