missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-1-methylpyrrolidinium bromide, 98%, Thermo Scientific Chemicals

Catalog Number p-7056877
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
69227-51-6
C7H16BrN
194.116
MFCD03095384
KHJQQUGSPDBDRM-UHFFFAOYSA-M
1-ethyl-1-methylpyrrolidinium bromide, 1-ethyl-1-methylpyrrolidin-1-ium bromide, 1-ethyl-1-methylpyrrolidiniumbromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide, 1-methyl-1-ethylpyrrolidinium bromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide 1:1, dsstox_cid_29306, dsstox_rid_83422, dsstox_gsid_49347, ksc495s4d
112264
1-ethyl-1-methylpyrrolidin-1-ium;bromide
CC[N+]1(CCCC1)C.[Br-]
This item is not returnable. View return policy
69227-51-6
C7H16BrN
194.116
MFCD03095384
KHJQQUGSPDBDRM-UHFFFAOYSA-M
1-ethyl-1-methylpyrrolidinium bromide, 1-ethyl-1-methylpyrrolidin-1-ium bromide, 1-ethyl-1-methylpyrrolidiniumbromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide, 1-methyl-1-ethylpyrrolidinium bromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide 1:1, dsstox_cid_29306, dsstox_rid_83422, dsstox_gsid_49347, ksc495s4d
112264
1-ethyl-1-methylpyrrolidin-1-ium;bromide
CC[N+]1(CCCC1)C.[Br-]

1-Ethyl-1-methylpyrrolidinium bromide s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethyl-1-methylpyrrolidinium bromide s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY
Quantity 25 g
Beilstein 14507266
Solubility Information Soluble in water.
Formula Weight 194.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-Ethyl-1-methylpyrrolidinium bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'