missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium bromide, 98+%, Thermo Scientific Chemicals

Catalog Number p-7047793
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
65039-08-9
C6H11BrN2
191.07
MFCD03427610
GWQYPLXGJIXMMV-UHFFFAOYSA-M
1-ethyl-3-methylimidazolium bromide, 3-ethyl-1-methyl-1h-imidazol-3-ium bromide, 1-ethyl-3-methylmidazolium bromide, unii-xo254ye73i, 1-ethyl-3-methyl-1h-imidazol-3-ium bromide, acmc-1b3a0, dsstox_cid_29300, dsstox_rid_83418, dsstox_gsid_49343, c6h11n2.br
2734235
1-ethyl-3-methylimidazol-3-ium;bromide
[Br-].CCN1C=C[N+](C)=C1
This item is not returnable. View return policy
65039-08-9
C6H11BrN2
191.07
MFCD03427610
GWQYPLXGJIXMMV-UHFFFAOYSA-M
1-ethyl-3-methylimidazolium bromide, 3-ethyl-1-methyl-1h-imidazol-3-ium bromide, 1-ethyl-3-methylmidazolium bromide, unii-xo254ye73i, 1-ethyl-3-methyl-1h-imidazol-3-ium bromide, acmc-1b3a0, dsstox_cid_29300, dsstox_rid_83418, dsstox_gsid_49343, c6h11n2.br
2734235
1-ethyl-3-methylimidazol-3-ium;bromide
[Br-].CCN1C=C[N+](C)=C1

An ionic liquid, 1-ethyl-3-methylimidazolium bromide is used as reaction medium for the synthesis and crystallization of a coordination polymer, (EMI)[Cd(BTC)]. 1-ethyl-3-methylimidazolium bromide was used as a solvent in [emim] 2 [Cd 2 (btec)Br 2 ]. Form the diene complexation; carbonyl insertion; cycloaddition; reduction and deoxygenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An ionic liquid, 1-ethyl-3-methylimidazolium bromide is used as reaction medium for the synthesis and crystallization of a coordination polymer, (EMI)[Cd(BTC)]. 1-ethyl-3-methylimidazolium bromide was used as a solvent in [emim] 2 [Cd 2 (btec)Br 2 ]. Form the diene complexation; carbonyl insertion; cycloaddition; reduction and deoxygenation.

Solubility
Soluble in water, dichloromethane, ethanol, acetonitrile. Insoluble in ethyl acetate, ether, and alkane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY
Melting Point 70°C to 73°C
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,dichloromethane,ethanol,acetonitrile. Insoluble in ethyl acetate,ether,and alkane.
Formula Weight 191.07
Percent Purity ≥98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'