Learn More
1-Ethyl-3-methylimidazolium diethyl phosphate, 98%, Thermo Scientific Chemicals
Catalog Number p-7029884
Click to view available options
Quantity:
5 g
25 g
848641-69-0
C10H21N2O4P
264.26
MFCD09953486
HQWOEDCLDNFWEV-UHFFFAOYSA-M
1-ethyl-3-methylimidazolium diethyl phosphate, 1-ethyl-3-methylimidazolium diethylphosphate, dsstox_cid_27865, dsstox_rid_82618, dsstox_gsid_47889, emim dep, basionics™ lq 11, 1-ethyl-3-methylimidazolium-diethyl phosphate, 1-ethyl-3-methylimidazolium diethyl phosphate hplc
53316406
diethyl phosphate;1-ethyl-3-methylimidazol-3-ium
CCN1C=C[N+](C)=C1.CCOP([O-])(=O)OCC
848641-69-0
C10H21N2O4P
264.26
MFCD09953486
HQWOEDCLDNFWEV-UHFFFAOYSA-M
1-ethyl-3-methylimidazolium diethyl phosphate, 1-ethyl-3-methylimidazolium diethylphosphate, dsstox_cid_27865, dsstox_rid_82618, dsstox_gsid_47889, emim dep, basionics™ lq 11, 1-ethyl-3-methylimidazolium-diethyl phosphate, 1-ethyl-3-methylimidazolium diethyl phosphate hplc
53316406
diethyl phosphate;1-ethyl-3-methylimidazol-3-ium
CCN1C=C[N+](C)=C1.CCOP([O-])(=O)OCC
It finds its application in improving enzymatic hydrolysis of wheat straw and in the enzymatic in situ saccharification of cellulose in aqueous-ionic liquid media.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application in improving enzymatic hydrolysis of wheat straw and in the enzymatic in situ saccharification of cellulose in aqueous-ionic liquid media.
Solubility
Fully miscible in water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY
Melting Point 19°C to 21°C
Density 1.157
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Fully miscible in water.
Formula Weight 264.26
Percent Purity 98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium diethyl phosphate