missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-4-fluorobenzene, 97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1-Ethyl-4-fluorobenzene is used as pharmaceutical Intermediates.
Solubility
Not miscible in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
1-Ethyl-4-fluorobenzene is used as pharmaceutical Intermediates.
Solubility
Not miscible in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
Specifications
Boiling Point | 139°C |
Flash Point | 30°C (86°F) |
Refractive Index | 1.471 |
Quantity | 5 g |
UN Number | UN1993 |
Beilstein | 1856987 |
Solubility Information | Not miscible in water. |
Formula Weight | 124.16 |
Percent Purity | 97% |
Chemical Name or Material | 1-Ethyl-4-fluorobenzene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'