Läs mer
1-Ethyl-6-fluoro-7-(piperazin-1-yl)quinolin-4(1H)-one, Mikromol™
Produktkod. 30051893
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051893 25 mg
17983006 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051893 Leverantör Mikromol Leverantörsnummer MM0350.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Ethyl-6-fluoro-7-(piperazin-1-yl)quinolin-4(1H)-one
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Ethyl-6-fluoro-7-(piperazin-1-yl)quinolin-4(1H)-one
CAS 75001-82-0
Analyt- eller komponentnamn 1-Ethyl-6-fluoro-7-(piperazin-1-yl)quinolin-4(1H)-one
Frakt skick Room Temperature
Molekylformel C15 H18 F N3 O
Certifikat och efterlevnad ISO 17025
Synonym 1-Ethyl-6-fluoro-7-(piperazin-1-yl)quinolin-4(1H)-one, Norfloxacin Imp. D (EP), Norfloxacin Impurity D
InChI formel InChI=1S/C15H18FN3O/c1-2-18-6-3-15(20)11-9-12(16)14(10-13(11)18)19-7-4-17-5-8-19/h3,6,9-10,17H,2,4-5,7-8H2,1H3
LEDER CCN1C=CC(=O)c2cc(F)c(cc12)N3CCNCC3
IUPAC-namn 1-ethyl-6-fluoro-7-piperazin-1-ylquinolin-4-one
Molekylvikt (g/mol) 275.32
Kvantitet 25 mg
Formel vikt 275.1434 g/mol