missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals

Catalog Number p-7081753
Click to view available options
Quantity:
5 g
25 g
100 g
70-34-8
C6H3FN2O4
186.098
MFCD00007056
LOTKRQAVGJMPNV-UHFFFAOYSA-N
2,4-dinitrofluorobenzene, dinitrofluorobenzene, fdnb, dnfb, 2,4-dinitro-1-fluorobenzene, sanger's reagent, fluoro-2,4-dinitrobenzene, 2,4-dinitrobenzenefluoride, 2,4-dinitrophenyl fluoride, fluorodinitrobenzene
6264
CHEBI:53049
1-fluoro-2,4-dinitrobenzene
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F
70-34-8
C6H3FN2O4
186.098
MFCD00007056
LOTKRQAVGJMPNV-UHFFFAOYSA-N
2,4-dinitrofluorobenzene, dinitrofluorobenzene, fdnb, dnfb, 2,4-dinitro-1-fluorobenzene, sanger's reagent, fluoro-2,4-dinitrobenzene, 2,4-dinitrobenzenefluoride, 2,4-dinitrophenyl fluoride, fluorodinitrobenzene
6264
CHEBI:53049
1-fluoro-2,4-dinitrobenzene
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F

1-Fluoro-2,4-dinitrobenzene is used to identify the amino acid sequence. It reacts with amino group of amino acids to yield dinitrophenyl-amino acids. It is also used in chromatographic methods. Further, it acts as an alkylating agent used in elucidating amino acid sequence in proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoro-2,4-dinitrobenzene is used to identify the amino acid sequence. It reacts with amino group of amino acids to yield dinitrophenyl-amino acids. It is also used in chromatographic methods. Further, it acts as an alkylating agent used in elucidating amino acid sequence in proteins.

Solubility
Soluble in chloroform, benzene, ether and propylene glycol.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Melting Point 24°C to 30°C
Density 1.482
Boiling Point 296°C
Flash Point >110°C (230°F)
Refractive Index 1.569
Quantity 5 g
UN Number UN2811
Beilstein 398632
Merck Index 14,4172
Solubility Information Soluble in chloroform,benzene,ether and propylene glycol.
Formula Weight 186.1
Percent Purity 99%
Chemical Name or Material 1-Fluoro-2,4-dinitrobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'