Learn More
1-Fluoro-4-(trifluoromethoxy)benzene, 99%, Thermo Scientific™
Catalog Number p-7024903
Click to view available options
Quantity:
1 g
5 g
352-67-0
C7H4F4O
180.102
MFCD00040835
JULMJGDXANEQDP-UHFFFAOYSA-N
1-fluoro-4-trifluoromethoxy benzene, 4-trifluoromethoxy fluorobenzene, 4-fluoro trifluoromethoxy benzene, benzene, 1-fluoro-4-trifluoromethoxy, p-fluoro trifluoromethoxy benzene, 4-trifluoromethoxy-1-fluorobenzene, p,alpha,alpha,alpha-tetrafluoroanisole, 4-fluorotrifluoromethoxybenzene, p-fluorophenyl trifluoromethyl ether
67698
1-fluoro-4-(trifluoromethoxy)benzene
C1=CC(=CC=C1OC(F)(F)F)F
352-67-0
C7H4F4O
180.102
MFCD00040835
JULMJGDXANEQDP-UHFFFAOYSA-N
1-fluoro-4-trifluoromethoxy benzene, 4-trifluoromethoxy fluorobenzene, 4-fluoro trifluoromethoxy benzene, benzene, 1-fluoro-4-trifluoromethoxy, p-fluoro trifluoromethoxy benzene, 4-trifluoromethoxy-1-fluorobenzene, p,alpha,alpha,alpha-tetrafluoroanisole, 4-fluorotrifluoromethoxybenzene, p-fluorophenyl trifluoromethyl ether
67698
1-fluoro-4-(trifluoromethoxy)benzene
C1=CC(=CC=C1OC(F)(F)F)F
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is employed as an active pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Density 1.323
Boiling Point 104°C to 105°C
Flash Point 15°C (59°F)
Odor Characteristic
Refractive Index 1.394
Quantity 1 g
UN Number UN1993
Beilstein 2046330
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 180.1
Percent Purity 99%
Chemical Name or Material 1-Fluoro-4-(trifluoromethoxy)benzene