Läs mer
1-Formylcyclohexanecarboxylic Acid Methyl Ester, TRC
Produktkod. 30366058
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366058 10 mg
30359357 100 mg
30356917 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366058 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC99893010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 84393-05-5
Molekylformel C9H14O3
Molekylvikt (g/mol) 170.21
Synonym Methyl 1-formylcyclohexane-1-carboxylate
IUPAC-namn methyl 1-formylcyclohexane-1-carboxylate
LEDER COC(=O)C1(CCCCC1)C=O
Specifikationer
Kemiskt namn eller material 1-Formylcyclohexanecarboxylic Acid Methyl Ester
Kvantitet 10 mg
InChI formel InChI=1S/C9H14O3/c1-12-8(11)9(7-10)5-3-2-4-6-9/h7H,2-6H2,1H3
Formel vikt 170.0943