Läs mer
1-heptanesulfinic Acid Sodium Salt, TRC
Produktkod. 30012474
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012474 25 mg
30009402 100 mg
30007432 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012474 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP27030525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68555-66-8
Molekylformel C7F15NaO2S
Molekylvikt (g/mol) 456.1
Synonym Perfluoroheptanesulfonate Na Salt;
IUPAC-namn sodium;1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptane-1-sulfinate
LEDER [Na+].[O-]S(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifikationer
Kemiskt namn eller material 1-heptanesulfinic Acid Sodium Salt
Kvantitet 25 mg
InChI formel InChI=1S/C7HF15O2S.Na/c8-1(9,2(10,11)4(14,15)6(18,19)20)3(12,13)5(16,17)7(21,22)25(23)24;/h(H,23,24);/q;+1/p-1
Formel vikt 455.9277