missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-Hexyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
(1-Hexyl)triphenylphosphonium bromide can be used as catalyst in pharmaceutical research. It is used in organic synthesis. Reactive wittg reagent used in prostaglandin synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(1-Hexyl)triphenylphosphonium bromide can be used as catalyst in pharmaceutical research. It is used in organic synthesis. Reactive wittg reagent used in prostaglandin synthesis.
Solubility
Soluble in water and methanol.
Notes
Hygroscopic. Store under inert gas. Protect from light. Store away from oxidizing materials.
(1-Hexyl)triphenylphosphonium bromide can be used as catalyst in pharmaceutical research. It is used in organic synthesis. Reactive wittg reagent used in prostaglandin synthesis.
Solubility
Soluble in water and methanol.
Notes
Hygroscopic. Store under inert gas. Protect from light. Store away from oxidizing materials.
Specifications
Melting Point | 199°C to 202°C |
Quantity | 25 g |
Beilstein | 3580731 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water and methanol. |
IUPAC Name | hexyltriphenylphosphanium bromide |
Formula Weight | 427.37 |
Percent Purity | 98% |
Chemical Name or Material | (1-Hexyl)triphenylphosphonium bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'