Learn More
1-Hexyne, Sigma-Aldrich™
Product Code. 15821091
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15821091 100 mL 100mL
15811091 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15821091 Supplier Sigma Aldrich Supplier No. 8206430100
 Restricted Item
Beräknad leverans: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 693-02-7
Grade Research
Molecular Weight (g/mol) 82.14
Specifications
Melting Point -132°C
Density 0.72 g/cm3 at 20°C
Boiling Point 71°C to 72°C/1013 hPa
MDL Number MFCD00009504
Quantity 100 mL
Synonym 1-Hexyne, 1-Hexyne