Läs mer
1’-Hydroxy Bufuralol (Mixture of Diastereomers), TRC
Produktkod. 17980785
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17980785 10 mg
17932595 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17980785 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH83048010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57704-16-2
Molekylformel C16 H23 N O3
Molekylvikt (g/mol) 277.36
Synonym 1-Hydroxy Bufuralol (Mixture Of Diastereomers)
IUPAC-namn 2-(tert-butylamino)-1-[7-(1-hydroxyethyl)-1-benzofuran-2-yl]ethanol
LEDER CC(O)c1cccc2cc(oc12)C(O)CNC(C)(C)C
Specifikationer
Kemiskt namn eller material 1'-Hydroxybufuralol
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C16H23NO3/c1-10(18)12-7-5-6-11-8-14(20-15(11)12)13(19)9-17-16(2,3)4/h5-8,10,13,17-19H,9H2,1-4H3
Formel vikt 277.1678
Procent renhet >95%