Läs mer
1-Hydroxycyclopentyl 2-chlorophenyl Ketone, TRC
Produktkod. 30136838
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136838 250 mg
30134183 100 mg
30006129 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136838 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH949885250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 90717-17-2
Molekylformel C12 H13 Cl O2
Molekylvikt (g/mol) 224.68
Synonym (2-Chlorophenyl)(1-hydroxycyclopentyl)methanone, 1-Hydroxycyclopentyl 2-chlorophenyl ketone, o-Chlorophenyl 1-hydroxycyclopentyl ketone
IUPAC-namn (2-chlorophenyl)-(1-hydroxycyclopentyl)methanone
LEDER OC1(CCCC1)C(=O)c2ccccc2Cl
Specifikationer
Kemiskt namn eller material (2-Chlorophenyl)(1-hydroxycyclopentyl)methanone
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C12H13ClO2/c13-10-6-2-1-5-9(10)11(14)12(15)7-3-4-8-12/h1-2,5-6,15H,3-4,7-8H2
Formel vikt 224.0604
Procent renhet >95%