Learn More
1-Hydroxyisoquinoline, 98%, BLD Pharm™
Product Code. 31006321
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
250mg
25g
500g
5g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31006321 10 g 10g
31002933 100 g 100g
30989972 1 g 1g
30994813 25 g 25g
31006038 250 mg 250mg
30994819 500 g 500g
30999689 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 31006321 Supplier BLD Pharm Supplier No. BD1157810g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 491-30-5
Molecular Formula C9H7NO
MDL Number MFCD00006899
Quantity 10 g
SMILES C1=CC=CC2=C1C(=O)NC=C2
Molecular Weight (g/mol) 145.16
Percent Purity 98%
Chemical Name or Material 1-Hydroxyisoquinoline