missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxypyrene, 99+%, Thermo Scientific Chemicals

Catalog Number p-147245
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5315-79-7
C16H10O
218.25
MFCD00044543
BIJNHUAPTJVVNQ-UHFFFAOYSA-N
1-hydroxypyrene, 1-pyrenol, 3-hydroxypyrene, pyrenol, unii-n2h6o5v707, hydroxypyrene, 1-hydroxy-pyrene, 3-pyrenol, 1-pyrenol 8ci
21387
CHEBI:34093
pyren-1-ol
C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)O
This item is not returnable. View return policy
5315-79-7
C16H10O
218.25
MFCD00044543
BIJNHUAPTJVVNQ-UHFFFAOYSA-N
1-hydroxypyrene, 1-pyrenol, 3-hydroxypyrene, pyrenol, unii-n2h6o5v707, hydroxypyrene, 1-hydroxy-pyrene, 3-pyrenol, 1-pyrenol 8ci
21387
CHEBI:34093
pyren-1-ol
C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 176°C to 178°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 50 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in most common organic solvents
Formula Weight 218.25
Percent Purity 99+%
Physical Form Powder or Fine Needles
Chemical Name or Material 1-Hydroxypyrene, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Hydroxypyrene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'