Läs mer
1-Hydroxypyridine-2-thione Zinc, TRC
Produktkod. 30132994
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132994 5 g
30135750 10 g
18073230 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132994 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH2498205G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13463-41-7
Molekylformel 2 C5 H4 N O S . Zn
Molekylvikt (g/mol) 317.72
Synonym Zinc, bis(1-hydroxy-2(1H)-pyridinethionato)- (6CI,7CI,8CI), Zinc, bis(1-hydroxy-2(1H)-pyridinethionato-O,S)-, (T-4)-, (T-4)-Bis[1-(hydroxy-κO)-2(1H)-pyridinethionato-κS2]zinc, 2(1H)-Pyridinethione, 1-hydroxy-, zinc complex, 1-Hydroxy-2-pyridinethione, zinc salt, 2-Mercaptopyridine 1-oxide zinc salt, 2-Pyridinethiol 1-oxide zinc salt, 2-Pyridinethiol N-oxide zinc salt, 2-Pyridylthiol-1-oxide zinc salt, AF-Z, BC-J, Biocut ZP, Bis(1-hydroxy-2(1H)-pyridinethionato)zinc, Bis(1-hydroxy-2-(1H)-pyridinethionato)zinc, Bis(2-pyridinethiol 1-oxide)zinc, Clean-Bio ZP, Danex, Dantrol, Denistat ANK, Desquaman, Evafine P 50, FSB 8332, Finecide ZPT, Hokucide ZPT, Hybricide 89, Intercide ZNP, Marukacide YP-DP, Microban Additive ZO 1, Microban Z 01, NSC 290409, Niccanon SKT, Niccanon ZP, OM 1563, Omadine Zinc, Pyrithione zinc, SR-A 103, Sanaizol 200, Sanitized TH 22-27, Sanitized TH 27-24BT, Sebulon Shampoo, Slaoff 94, Slaoff 95, TH 22-27, Tomicide Z 50, Tomicide ZPT, Tomicide ZPT 100, Tomicide ZPT 50, Vancide P, Vedexil-PZ, ZNP Bar, ZO-E, ZOE-T, ZPT, Zinc 1-hydroxy-2-pyridinethione, Zinc 2-mercaptopyridine N-oxide, Zinc 2-pyridinethiol 1-oxide, Zinc Omadine, Zinc Omadine ZOE, Zinc Omadine ZOE Dispersion, Zinc Pyrion, Zinc bis(2-pyridylthio-1-oxide), Zinc pyrethion, Zinc pyridine-2-thione-N-oxide, Zinc pyrithione, Zinc, bis(2-pyridinylthio)-, N,N'-dioxide, Zincopan, Zincpolyanemine, Zn Omadine ZOE, ZnPT
IUPAC-namn zinc;1-oxidopyridin-1-ium-2-thiolate
LEDER [Zn+2].[O-][n+]1ccccc1[S-].[O-][n+]2ccccc2[S-]
Specifikationer
Kemiskt namn eller material Pyrithione Zinc
Kvantitet 5 g
InChI formel InChI=1S/2C5H5NOS.Zn/c2*7-6-4-2-1-3-5(6)8;/h2*1-4,8H;/q;;+2/p-2
Formel vikt 315.9319