missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Iodohexadecane, 98%, stab. with copper, Thermo Scientific™
408.20 SEK - 2320.00 SEK
Chemical Identifiers
CAS | 544-77-4 |
---|---|
Molecular Formula | C16H33I |
Molecular Weight (g/mol) | 352.344 |
MDL Number | MFCD00001089 |
InChI Key | KMWHQYDMBYABKL-UHFFFAOYSA-N |
Synonym | cetyl iodide, hexadecyl iodide, hexadecane, 1-iodo, hexadecane, iodo, unii-jo95x48m8o, 1-iodo-hexadecan, iodohexadecane, cetyljodid, 1-iodo-hexadecane, n-hexadecyl iodide |
PubChem CID | 11007 |
IUPAC Name | 1-iodohexadecane |
SMILES | CCCCCCCCCCCCCCCCI |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15423067
|
Thermo Scientific Alfa Aesar
A12977.14 |
25g |
408.20 SEK
25g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11399216
|
Thermo Scientific Alfa Aesar
A12977.22 |
100g |
1161.76 SEK
100g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11309226
|
Thermo Scientific Alfa Aesar
A12977.30 |
250g |
2320.00 SEK
250g |
Estimated Shipment: 06-07-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
1-Iodohexadecane, is used as an ionic liquid for electrochemical applications. It is also used in detection methods as primer and probe design instrumentation applications guide in qPCR Technic.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications1-Iodohexadecane, is used as an ionic liquid for electrochemical applications. It is also used in detection methods as primer and probe design instrumentation applications guide in qPCR Technic.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away from strong oxidizing agents, strong bases. Store it at a temperature of 15 - 30°C. Stable under normal temperatures and pressures.
Chemical Identifiers
544-77-4 | |
352.344 | |
KMWHQYDMBYABKL-UHFFFAOYSA-N | |
11007 | |
CCCCCCCCCCCCCCCCI |
C16H33I | |
MFCD00001089 | |
cetyl iodide, hexadecyl iodide, hexadecane, 1-iodo, hexadecane, iodo, unii-jo95x48m8o, 1-iodo-hexadecan, iodohexadecane, cetyljodid, 1-iodo-hexadecane, n-hexadecyl iodide | |
1-iodohexadecane |
Specifications
1-Iodohexadecane | |
544-77-4 | |
21°C to 23°C | |
>110°C (230°F) | |
CH3(CH2)15I | |
1748533 | |
cetyl iodide, hexadecyl iodide, hexadecane, 1-iodo, hexadecane, iodo, unii-jo95x48m8o, 1-iodo-hexadecan, iodohexadecane, cetyljodid, 1-iodo-hexadecane, n-hexadecyl iodide | |
Insoluble in water. | |
KMWHQYDMBYABKL-UHFFFAOYSA-N | |
1-iodohexadecane | |
11007 | |
98% |
stabilized with copper | |
1.121 | |
206°C to 207°C (10mmHg) | |
C16H33I | |
MFCD00001089 | |
1.482 | |
25g | |
Light sensitive | |
CCCCCCCCCCCCCCCCI | |
352.344 | |
352.34 |
RUO – Research Use Only