Learn More
1-Iodooctane, 98+%, stab. with copper, Thermo Scientific™
300.78 SEK - 2562.39 SEK
Chemical Identifiers
CAS 629-27-6
Molecular Formula C8H17I
Molecular Weight (g/mol) 240.128
MDL Number MFCD00001105
InChI Key UWLHSHAHTBJTBA-UHFFFAOYSA-N
Synonym n-octyl iodide, octyl iodide, octane, 1-iodo, 1-octyl iodide, 1-n-octyl iodide, 1-jodoktan, 1-jodoktan czech, 1-iodooctane;n-octyl iodide, iodoctane, iodooctane
PubChem CID 12380
IUPAC Name 1-iodooctane
SMILES CCCCCCCCI
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11405846
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
B22214.14
25g
300.78 SEK
25g
View alternative products
Save Up to 
»
In Stock
to see stock.
11415846
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
B22214.22
100g
756.80 SEK
100g
View alternative products
Save Up to 
»
In Stock
to see stock.
11425846
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
B22214.36
500g
2562.39 SEK
500g
View alternative products
Save Up to 
»
In Stock
to see stock.
Description
Description
Alkylation of [2,2?-([2,2?-bipyridine]-6,6?-diyl)bis[phenolato]-N,N?,O,O?]nickel(II)was performed using the catalytic reduction of 1-iodooctane. Primary alkyl iodide 1-iodooctane reacts with lithium aluminum deuteride (LAD) in a 1:0.2 ratio to form 1-deuteriooctane.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Alkylation of [2,2′-([2,2′-bipyridine]-6,6′-diyl)bis[phenolato]-N,N′,O,O′]nickel(II)was performed using the catalytic reduction of 1-iodooctane. Primary alkyl iodide 1-iodooctane reacts with lithium aluminum deuteride (LAD) in a 1:0.2 ratio to form 1-deuteriooctane.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, bases.
Specifications
Chemical Identifiers
629-27-6
240.128
UWLHSHAHTBJTBA-UHFFFAOYSA-N
12380
CCCCCCCCI
C8H17I
MFCD00001105
n-octyl iodide, octyl iodide, octane, 1-iodo, 1-octyl iodide, 1-n-octyl iodide, 1-jodoktan, 1-jodoktan czech, 1-iodooctane;n-octyl iodide, iodoctane, iodooctane
1-iodooctane
Specifications
1-Iodooctane
629-27-6
-46°C
95°C (203°F)
CH3(CH2)7I
1697479
n-octyl iodide, octyl iodide, octane, 1-iodo, 1-octyl iodide, 1-n-octyl iodide, 1-jodoktan, 1-jodoktan czech, 1-iodooctane;n-octyl iodide, iodoctane, iodooctane
Insoluble in water.
UWLHSHAHTBJTBA-UHFFFAOYSA-N
1-iodooctane
12380
≥98%
stabilized with copper
1.325
225°C to 226°C
C8H17I
MFCD00001105
1.4878
25g
Light sensitive
CCCCCCCCI
240.128
240.13