missing translation for 'onlineSavingsMsg'
Learn More

1-Iodooctane, 98%, stabilized, Thermo Scientific Chemicals

Catalog Number p-4258874
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
629-27-6
C8H17I
240.13
UWLHSHAHTBJTBA-UHFFFAOYSA-N
n-octyl iodide, octyl iodide, octane, 1-iodo, 1-octyl iodide, 1-n-octyl iodide, 1-jodoktan, 1-jodoktan czech, 1-iodooctane;n-octyl iodide, iodoctane, iodooctane
12380
1-iodooctane
CCCCCCCCI
This item is not returnable. View return policy
629-27-6
C8H17I
240.13
UWLHSHAHTBJTBA-UHFFFAOYSA-N
n-octyl iodide, octyl iodide, octane, 1-iodo, 1-octyl iodide, 1-n-octyl iodide, 1-jodoktan, 1-jodoktan czech, 1-iodooctane;n-octyl iodide, iodoctane, iodooctane
12380
1-iodooctane
CCCCCCCCI

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -46°C to 45°C
Color Colorless to Amber
Density 1.3300g/mL
Boiling Point 225.0°C to 226.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7I
Refractive Index 1.4878
Quantity 100 g
Beilstein 01,16
Specific Gravity 1.33
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most common organic solvents
Stabilizer copper
Formula Weight 240.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Iodooctane, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • 1-Iodooctane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'