Learn More
1-Iodopentane, 98%, stab with copper, Thermo Scientific Chemicals
Catalog Number p-7025349
Click to view available options
Quantity:
100 g
500 g
628-17-1
C5H11I
198.047
MFCD00001100
BLXSFCHWMBESKV-UHFFFAOYSA-N
n-amyl iodide, amyl iodide, pentyl iodide, n-pentyl iodide, pentane, 1-iodo, 1-pentyl iodide, 1-jodpentan, 1-jodpentan czech, iodopentane, pentyliodide
12335
1-iodopentane
CCCCCI
628-17-1
C5H11I
198.047
MFCD00001100
BLXSFCHWMBESKV-UHFFFAOYSA-N
n-amyl iodide, amyl iodide, pentyl iodide, n-pentyl iodide, pentane, 1-iodo, 1-pentyl iodide, 1-jodpentan, 1-jodpentan czech, iodopentane, pentyliodide
12335
1-iodopentane
CCCCCI
1-Iodopentane (pentyl iodide) was used in the preparation of 1-pentyl radical by shock heating process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Iodopentane (pentyl iodide) was used in the preparation of 1-pentyl radical by shock heating process.
Solubility
Insoluble in water.
Notes
Light sensitive. Store in dark place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -86°C
Density 1.515
Boiling Point 154°C to 156°C
Flash Point 51°C (123°F)
Odor Characteristic
Linear Formula CH3(CH2)4I
Refractive Index 1.495
Quantity 100 g
UN Number UN1993
Beilstein 1730982
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 198.05
Percent Purity 98%
Chemical Name or Material 1-Iodopentane, stabilized with copper