missing translation for 'onlineSavingsMsg'
Learn More

1-Iodoperfluoro(4-methyl-2-pentene), 94%, Thermo Scientific™

Catalog Number 15459049
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 120695-78-5
Boiling Point 32°C to 33°C (50 mmHg)
Molecular Formula C6F11I
MDL Number MFCD00153199
Quantity 5 g
Beilstein 3618405
Sensitivity Light sensitive
Synonym 1,1,2,3,4,5,5,5-octafluoro-1-iodo-4-trifluoromethyl pent-2-ene, 2-pentene,1,1,2,3,4,5,5,5-octafluoro-1-iodo-4-trifluoromethyl-, e-9ci, 1-iodoperfluoro 4-methyl-2-pentene, acmc-20mp2k, 1,1,2,3,4,5,5,5-octafluoro-1-iodo-4-trifluoromethyl-2-pentene, 1,1,2,3,4,5,5,5-octakis fluoranyl-1-iodanyl-4-trifluoromethyl pent-2-ene
InChI Key BJZGOHYMGKSXBU-UHFFFAOYSA-N
SMILES C(=C(C(F)(F)I)F)(C(C(F)(F)F)(C(F)(F)F)F)F
IUPAC Name 1,1,2,3,4,5,5,5-octafluoro-1-iodo-4-(trifluoromethyl)pent-2-ene
Molecular Weight (g/mol) 407.953
PubChem CID 53395385
Formula Weight 407.95
Percent Purity 94%
Chemical Name or Material 1-Iodoperfluoro(4-methyl-2-pentene)
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'