Läs mer
1-Isobutyl-1H-imidazo[4,5-c]quinoline 5-Oxide (Desaminoimiquimod N-Oxide), Mikromol™
Produktkod. 30052626
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052626 25 mg
30051547 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30052626 Leverantör Mikromol Leverantörsnummer MM1158.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Isobutyl-1H-imidazo[4,5-c]quinoline 5-Oxide (Desaminoimiquimod N-Oxide)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desaminoimiquimod N-Oxide
CAS 99010-63-6
Analyt- eller komponentnamn Desaminoimiquimod N-Oxide
Frakt skick Room Temperature
Molekylformel C14 H15 N3 O
Certifikat och efterlevnad ISO 9001
Synonym 1-Isobutyl-1H-imidazo[4,5-c]quinoline 5-oxide, Imiquimod Related Compound B
InChI formel InChI=1S/C14H15N3O/c1-10(2)7-16-9-15-12-8-17(18)13-6-4-3-5-11(13)14(12)16/h3-6,8-10H,7H2,1-2H3
LEDER CC(C)Cn1cnc2c[n+]([O-])c3ccccc3c12
IUPAC-namn 1-(2-methylpropyl)-5-oxidoimidazo[4,5-c]quinolin-5-ium
Molekylvikt (g/mol) 241.29
Kvantitet 25 mg
Formel vikt 241.1215 g/mol