Läs mer
1-Isopropylamino-3-phenoxypropan-2-ol, TRC
Produktkod. 17903175
Change view
Kvantitet:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17903175 5 g
17993275 1 g
17952865 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17903175 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI8240705G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7695-63-8
Molekylformel C12 H19 N O2
Molekylvikt (g/mol) 209.28
Synonym (2RS)-1-[(1-Methylethyl)amino]-3-phenoxypropan-2-ol, Metoprolol Succinate Imp. F (EP), Metoprolol Tartrate Imp. F (EP)
IUPAC-namn 1-phenoxy-3-(propan-2-ylamino)propan-2-ol
LEDER CC(C)NCC(O)COc1ccccc1
Specifikationer
Kemiskt namn eller material (2RS)-1-[(1-Methylethyl)amino]-3-phenoxy-propan-2-ol
Kvantitet 5 g
InChI formel InChI=1S/C12H19NO2/c1-10(2)13-8-11(14)9-15-12-6-4-3-5-7-12/h3-7,10-11,13-14H,8-9H2,1-2H3
Formel vikt 209.1416