Läs mer
1-Kestose, TRC
Produktkod. 30406104
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30406104 100 mg
1 options
Produktkod. 30406104 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK264850100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Kestose
CAS 470-69-9
Molekylformel C18 H32 O16
Kvantitet 100 mg
Synonym Glucopyranoside, O-β-D-fructofuranosyl-(2→1)-β-D-fructofuranosyl, α-D- (7CI,8CI), α-D-Glucopyranoside, O-β-D-fructofuranosyl-(2→1)-O-β-D-fructofuranosyl-(2→1) (6CI), O-β-D-Fructofuranosyl-(2→1)-β-D-fructofuranosyl α-D-glucopyranoside, 1-Fructosylsucrose, 1-Kestose, 1-Kestotriose, 1F-Fructosylsucrose, GF 2
InChI formel InChI=1S/C18H32O16/c19-1-6-9(23)12(26)13(27)16(31-6)34-18(15(29)11(25)8(3-21)33-18)5-30-17(4-22)14(28)10(24)7(2-20)32-17/h6-16,19-29H,1-5H2/t6-,7-,8-,9-,10-,11-,12+,13-,14+,15+,16-,17-,18+/m1/s1
LEDER OC[C@H]1O[C@H](O[C@]2(CO[C@]3(CO)O[C@H](CO)[C@@H](O)[C@@H]3O)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC-namn (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-2-[[(2R,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxymethyl]-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molekylvikt (g/mol) 504.44
Formel vikt 504.169