Läs mer
1-Keto Ketorolac, TRC
Produktkod. 30135729
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135729 25 mg
18002340 100 mg
18012010 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135729 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK19530025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 113502-52-6
Molekylformel C14 H11 N O2
Molekylvikt (g/mol) 225.24
Synonym Ketorolac Trometamol Imp. B (EP), 5-Benzoyl-2,3-dihydro-1H-pyrrolizin-1-one, Ketorolac 1-Keto Analog
IUPAC-namn 5-benzoyl-2,3-dihydropyrrolizin-1-one
LEDER O=C(c1ccccc1)c2ccc3C(=O)CCn23
Specifikationer
Kemiskt namn eller material 5-Benzoyl-2,3-dihydro-1H-pyrrolizin-1-one
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C14H11NO2/c16-13-8-9-15-11(13)6-7-12(15)14(17)10-4-2-1-3-5-10/h1-7H,8-9H2
Formel vikt 225.079
Procent renhet >95%