Läs mer
1-Linoleoyl-(2S)-glycerol, TRC
Produktkod. 30471428
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30471428 500 mg
30477293 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30471428 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL467955500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 67968-46-1
Molekylformel C21H38O4
Molekylvikt (g/mol) 354.52
Synonym α-Linoleic Acid (2S)-2,3-Dihydroxypropyl Ester, (9Z,12Z)-9,12-Octadecadienoic Acid (2S)-2,3-Dihydroxypropyl Ester
IUPAC-namn [(2S)-2,3-dihydroxypropyl] (9Z,12Z)-octadeca-9,12-dienoate
LEDER CCCCC/C=C\C/C=C\CCCCCCCC(=O)OC[C@H](CO)O
Specifikationer
Kemiskt namn eller material 1-Linoleoyl-(2S)-glycerol
Kvantitet 500 mg
InChI formel InChI=1S/C21H38O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h6-7,9-10,20,22-23H,2-5,8,11-19H2,1H3/b7-6-,10-9-/t20-/m0/s1
Formel vikt 354.277