Läs mer
1-Methoxy-2-(methoxymethyl)-4-methyl-2-(propan-2-yl)pentane, TRC
Produktkod. 30137637
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137637 50 mg
30007519 10 mg
30009738 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137637 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM15611650MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 129228-21-3
Molekylformel C12H26O2
Molekylvikt (g/mol) 202.33
IUPAC-namn 3,3-bis(methoxymethyl)-2,5-dimethylhexane
LEDER COCC(COC)(CC(C)C)C(C)C
Specifikationer
Kemiskt namn eller material 1-Methoxy-2-(methoxymethyl)-4-methyl-2-(propan-2-yl)pentane
Kvantitet 50 mg
InChI formel InChI=1S/C12H26O2/c1-10(2)7-12(8-13-5,9-14-6)11(3)4/h10-11H,7-9H2,1-6H3
Formel vikt 202.1933