Läs mer
1-Methoxy-4-(2-nitrovinyl)benzene, TRC
Produktkod. 30179780
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179780 5 g
30179776 10 g
30180016 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179780 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3396985G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3179-10-0
Molekylformel C9H9NO3
Molekylvikt (g/mol) 179.17
IUPAC-namn 1-methoxy-4-(2-nitroethenyl)benzene
LEDER COc1ccc(C=C[N+](=O)[O-])cc1
Specifikationer
Kemiskt namn eller material 1-Methoxy-4-(2-nitrovinyl)benzene
Kvantitet 5 g
InChI formel InChI=1S/C9H9NO3/c1-13-9-4-2-8(3-5-9)6-7-10(11)12/h2-7H,1H3
Formel vikt 179.0582