missing translation for 'onlineSavingsMsg'
Läs mer

1-Methoxy-4-nitro-2-phenoxybenzene, TRC

Product Code. 30472509
Change view
Click to view available options
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30472509 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30472509 Supplier Toronto Research Chemicals Supplier No. TRCM49696550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 1-Methoxy-4-nitro-2-phenoxybenzene
CAS 14064-93-8
Molekylformel C13 H11 N O4
Kvantitet 50 mg
InChI formel InChI=1S/C13H11NO4/c1-17-12-8-7-10(14(15)16)9-13(12)18-11-5-3-2-4-6-11/h2-9H,1H3
LEDER COc1ccc(cc1Oc2ccccc2)[N+](=O)[O-]
IUPAC-namn 1-methoxy-4-nitro-2-phenoxybenzene
Molekylvikt (g/mol) 245.231
Formel vikt 245.069
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.