Läs mer
1-Methyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one, Mikromol™
Produktkod. 30049835
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049835 25 mg
17921746 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049835 Leverantör Mikromol Leverantörsnummer MM0474.130025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Methyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Methyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one
CAS 1246819-06-6
Analyt- eller komponentnamn 1-Methyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one
Frakt skick Room Temperature
Molekylformel C14 H9 N3 O S
Certifikat och efterlevnad ISO 9001
Synonym 1-Methyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one
InChI formel InChI=1S/C14H9N3OS/c1-8-11(18)6-7-16-12(8)13(19)17-10-5-3-2-4-9(10)15-14(16)17/h2-7H,1H3
LEDER CC1=C2N(C=CC1=O)c3nc4ccccc4n3C2=S
IUPAC-namn 1-Methyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one
Molekylvikt (g/mol) 267.31
Kvantitet 25 mg
Formel vikt 267.0466 g/mol