missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-4-carbaldehyde, 97%, Thermo Scientific™

Catalog Number p-3484803
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
133994-99-7
C10H9NO
159.19
MFCD08690261
GFTNSZIYQMJJMD-UHFFFAOYSA-N
1-methyl-1h-indole-4-carbaldehyde, pubchem20444, acmc-20a8ho, 1-methylindole-4-carboxaldehyde, 1-methyl-1h-indol-4-carboxaldehyde, 1h-indole-4-carboxaldehyde, 1-methyl
17221111
1-methylindole-4-carbaldehyde
CN1C=CC2=C(C=O)C=CC=C12
This item is not returnable. View return policy
133994-99-7
C10H9NO
159.19
MFCD08690261
GFTNSZIYQMJJMD-UHFFFAOYSA-N
1-methyl-1h-indole-4-carbaldehyde, pubchem20444, acmc-20a8ho, 1-methylindole-4-carboxaldehyde, 1-methyl-1h-indol-4-carboxaldehyde, 1h-indole-4-carboxaldehyde, 1-methyl
17221111
1-methylindole-4-carbaldehyde
CN1C=CC2=C(C=O)C=CC=C12
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-4-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'