missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
1-Methyl-2,3,4,9-tetrahydro-1H-b-carboline-3-carboxylic Acid, TRC
Specifications
Specifications
| Kemiskt namn eller material | 1-Methyl-2,3,4,9-tetrahydro-1H-beta-carboline-3-carboxylic Acid |
| CAS | 5470-37-1 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C13H14N2O2 |
| Kvantitet | 100 mg |
| Synonym | 1H-Pyrido[3,4-b]indole-3-carboxylic acid, 2,3,4,9-tetrahydro-1-methyl-, 2,3,4,9-Tetrahydro-1-methyl-1H-pyrido[3,4-b]indole-3-carboxylic acid, 1,2,3,4-Tetrahydro-3-carboxyharmane, 1,2,3,4-Tetrahydroharman-3-carboxylic acid, 1-Methyl-1,2,3,4-Tetrahydropyrido[3,4-b]indole-3-carboxylic acid, 1-Methyl-1,2,3,4-tetrahydro-β-carboline-3-carboxylic acid, 1-Methyl-1H,2H,3H,4H,9H-pyrido[3,4-b]indole-3-carboxylic acid, 1-Methyl-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic acid, 1-Methyl-2,3,4,9-tetrahydro-1H-β-carboline-3-carboxylic acid, NSC 26225, Tetrahydroharman-3-carboxylic acid, 1-Methyl-1,2,3,4-tetrahydro-9H-β-carboline-3-carboxylic acid |
| InChI formel | InChI=1S/C13H14N2O2/c1-7-12-9(6-11(14-7)13(16)17)8-4-2-3-5-10(8)15-12/h2-5,7,11,14-15H,6H2,1H3,(H,16,17) |
| LEDER | CC1C2=C(CC(N1)C(=O)O)C3=CC=CC=C3N2 |
| IUPAC-namn | 1-methyl-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic acid |
| Molekylvikt (g/mol) | 230.26 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?