missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-phenylindole, 99%, Thermo Scientific Chemicals

Catalog Number p-7026220
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
3558-24-5
C15H13N
207.276
MFCD00022892
SFWZZSXCWQTORH-UHFFFAOYSA-N
1-methyl-2-phenyl-1h-indole, 2-phenyl-n-methylindole, 1h-indole, 1-methyl-2-phenyl, n-methyl-2-phenylindole, indole, 1-methyl-2-phenyl, pubchem7435, n-methyl-2-phenyl indole, n-methyl-2-phenyl-indole, acmc-1cp6x, 1-methyl-2-phenyl-indole
77095
1-methyl-2-phenylindole
CN1C2=CC=CC=C2C=C1C3=CC=CC=C3
This item is not returnable. View return policy
3558-24-5
C15H13N
207.276
MFCD00022892
SFWZZSXCWQTORH-UHFFFAOYSA-N
1-methyl-2-phenyl-1h-indole, 2-phenyl-n-methylindole, 1h-indole, 1-methyl-2-phenyl, n-methyl-2-phenylindole, indole, 1-methyl-2-phenyl, pubchem7435, n-methyl-2-phenyl indole, n-methyl-2-phenyl-indole, acmc-1cp6x, 1-methyl-2-phenyl-indole
77095
1-methyl-2-phenylindole
CN1C2=CC=CC=C2C=C1C3=CC=CC=C3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 98°C to 99°C
Quantity 10 g
Beilstein 148770
Formula Weight 207.28
Percent Purity 99%
Chemical Name or Material 1-Methyl-2-phenylindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'