missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog Number p-4812627
Click to view available options
Quantity:
1 L
2.5 L
872-50-4
C5H9NO
99.13
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methylpyrrolidin-2-one
CN1CCCC1=O
872-50-4
C5H9NO
99.13
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methylpyrrolidin-2-one
CN1CCCC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -24.0°C
Density 1.0300g/mL
Boiling Point 202.0°C
Quantity 1 L
Flash Point 91°C
Packaging Glass Bottle
Refractive Index 1.4690 to 1.4710
Assay Percent Range 99.4% min. (GC)
Specific Gravity 1.03
Viscosity 1.67 mPa.s (20°C)
Merck Index 15,619
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 99.13
Percent Purity 99.50%
Grade Analytical
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'