missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-3-piperidinemethanol, 95%, Thermo Scientific™
Click to view available options
Quantity:
50 g
250 g
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Density | 1.0100g/mL |
Boiling Point | 140.0°C to 145.0°C |
Flash Point | 94°C |
Assay Percent Range | 94% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.4772 to 1.4792 |
Quantity | 50 g |
Beilstein | 21,II,8 |
Show More |
Safety and Handling
Product Identifier
- 1-Methyl-3-piperidinemethanol
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'