Läs mer
1-Methyl-4,5,6,7-tetrahydro-1H-benzoimidazole, TRC
Produktkod. 30476487
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476487 500 mg
30466203 1 g
30472488 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30476487 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM341095500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1837-49-6
Molekylformel C8H12N2
Molekylvikt (g/mol) 136.2
IUPAC-namn 1-methyl-4,5,6,7-tetrahydrobenzimidazole
LEDER Cn1cnc2CCCCc12
Specifikationer
Kemiskt namn eller material 1-Methyl-4,5,6,7-tetrahydro-1H-benzoimidazole
Kvantitet 500 mg
InChI formel InChI=1S/C8H12N2/c1-10-6-9-7-4-2-3-5-8(7)10/h6H,2-5H2,1H3
Formel vikt 136.1