missing translation for 'onlineSavingsMsg'
Läs mer

1-Methyl-4-imidazoleacetic Acid Hydrochloride, TRC

Produktkod. 30133158
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133158 50 mg
30133838 100 mg
17942635 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30133158 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32380050MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35454-39-8
Molekylformel C6H9ClN2O2
Molekylvikt (g/mol) 176.6
Synonym 1-Methyl-1H-Imidazole-4-acetic Acid Monohydrochloride, 1-Methyl-4-imidazoleacetic Acid Hydrochloride, 2-(1-Methyl-1H-imidazol-4-yl)ethanoic Acid Hydrochloride
IUPAC-namn 2-(1-methylimidazol-4-yl)acetic acid;hydrochloride
LEDER C1(=CN(C)C=N1)CC(O)=O.Cl

Specifications

Kemiskt namn eller material 1-Methyl-4-imidazoleacetic Acid Hydrochloride
Kvantitet 50 mg
InChI formel InChI=1S/C6H8N2O2.ClH/c1-8-3-5(7-4-8)2-6(9)10;/h3-4H,2H2,1H3,(H,9,10);1H
Formel vikt 176.0353
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.